Läs mer
N-Boc-Lys-OtBu Hydrochloride, TRC
Produktkod. 30342170
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30342170 1 g
1 options
Produktkod. 30342170 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB6570651G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Boc-Lys-OtBu Hydrochloride
CAS 7750-45-0
Molekylformel C15H30N2O4 . HCl
Kvantitet 1 g
Synonym N2-[(1,1-Dimethylethoxy)carbonyl]-L-lysine 1,1-Dimethylethyl Ester Hydrochloride, N-Boc-lysine t-Butyl Ester Hydrochloride, Nalpha-(t-Butoxycarbonyl)lysine t-Butyl Ester Hydrochloride
InChI formel InChI=1S/C15H30N2O4.ClH/c1-14(2,3)20-12(18)11(9-7-8-10-16)17-13(19)21-15(4,5)6;/h11H,7-10,16H2,1-6H3,(H,17,19);1H/t11-;/m0./s1
LEDER Cl.CC(C)(C)OC(=O)N[C@@H](CCCCN)C(=O)OC(C)(C)C
IUPAC-namn tert-butyl (2S)-6-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoate;hydrochloride
Molekylvikt (g/mol) 338.87
Formel vikt 338.1972