Läs mer
N-Boc-Lysine Tert-butyl Ester, TRC
Produktkod. 30344780
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30344780 250 mg
1 options
Produktkod. 30344780 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB775045250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Boc-Lysine Tert-butyl Ester
CAS 7750-42-7
Molekylformel C15H30N2O4
Kvantitet 250 mg
Synonym Nalpha-(Tert-Butoxycarbonyl)lysine Tert-butyl Ester, (S)-tert-Butyl 6-amino-2-((tert-butoxycarbonyl)amino)hexanoate
InChI formel InChI=1S/C15H30N2O4/c1-14(2,3)20-12(18)11(9-7-8-10-16)17-13(19)21-15(4,5)6/h11H,7-10,16H2,1-6H3,(H,17,19)/t11-/m0/s1
LEDER CC(C)(C)OC(=O)N[C@@H](CCCCN)C(=O)OC(C)(C)C
IUPAC-namn tert-butyl (2S)-6-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoate
Molekylvikt (g/mol) 302.41
Formel vikt 302.2206