Learn More
Alfa Aesar™ N-Boc-N-methyl-L-tyrosine dicyclohexylammonium salt, 98%
Brand:  Alfa Aesar™ H63307.03
Additional Details : CAS Number : 95105-25-2 Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15498226
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Specifications
Chemical Identifiers
95105-25-2
476.658
DVADYPVUIPQNGV-YDALLXLXSA-N
46735204
CC(C)(C)OC(=O)N(C)C(CC1=CC=C(C=C1)O)C(=O)O.C1CCC(CC1)NC2CCCCC2
C27H44N2O5
MFCD00237569
boc-n-me-tyr-oh.dcha, boc-n-me-tyr-oh dcha, boc-n-me-tyr.dcha, dicyclohexylamine s-2-tert-butoxycarbonyl methyl amino-3-4-hydroxyphenyl propanoate, n-boc-n-methyl-l-tyrosine dicyclohexylammonium salt, pubchem20979, boc-n-me-tyr-ohcha, dicyclohexylamine s-2-boc methyl amino-3-4-hydroxyphenyl propanoate, n-alpha-t-butyloxycarbonyl-n-alpha-methyl-l-tyrosine dicyclohexylamine, 2s-2-tert-butoxycarbonyl methyl amino-3-4-hydroxyphenyl propanoic acid; dicha
N-cyclohexylcyclohexanamine;(2S)-3-(4-hydroxyphenyl)-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]propanoic acid
Specifications
N-Boc-N-methyl-L-tyrosine dicyclohexylammonium salt
C27H44N2O5
boc-n-me-tyr-oh.dcha, boc-n-me-tyr-oh dcha, boc-n-me-tyr.dcha, dicyclohexylamine s-2-tert-butoxycarbonyl methyl amino-3-4-hydroxyphenyl propanoate, n-boc-n-methyl-l-tyrosine dicyclohexylammonium salt, pubchem20979, boc-n-me-tyr-ohcha, dicyclohexylamine s-2-boc methyl amino-3-4-hydroxyphenyl propanoate, n-alpha-t-butyloxycarbonyl-n-alpha-methyl-l-tyrosine dicyclohexylamine, 2s-2-tert-butoxycarbonyl methyl amino-3-4-hydroxyphenyl propanoic acid; dicha
CC(C)(C)OC(=O)N(C)C(CC1=CC=C(C=C1)O)C(=O)O.C1CCC(CC1)NC2CCCCC2
476.658
476.65
98%
1g
95105-25-2
MFCD00237569
DVADYPVUIPQNGV-YDALLXLXSA-N
N-cyclohexylcyclohexanamine;(2S)-3-(4-hydroxyphenyl)-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]propanoic acid
46735204
Powder
White
Videos
SDS Documents