missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-N-methylethanolamine, 95%, Thermo Scientific Chemicals

Catalog Number p-7057218
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
57561-39-4
C8H17NO3
175.228
MFCD03425857
RFDSJHHLGFFVHD-UHFFFAOYSA-N
n-boc-n-methyl-aminoethanol, tert-butyl 2-hydroxyethyl methyl carbamate, boc,me-glycinol, tert-butyl n-2-hydroxyethyl-n-methylcarbamate, 2-n-methyl-n-boc-amino ethanol, 2-n-boc-n-methylamino ethanol, n-boc-n-methylethanolamine, 2-hydroxyethyl methylcarbamic acid 1,1-dimethylethyl ester, carbamic acid, n-2-hydroxyethyl-n-methyl-, 1,1-dimethylethyl ester
545700
tert-butyl N-(2-hydroxyethyl)-N-methylcarbamate
CC(C)(C)OC(=O)N(C)CCO
This item is not returnable. View return policy
57561-39-4
C8H17NO3
175.228
MFCD03425857
RFDSJHHLGFFVHD-UHFFFAOYSA-N
n-boc-n-methyl-aminoethanol, tert-butyl 2-hydroxyethyl methyl carbamate, boc,me-glycinol, tert-butyl n-2-hydroxyethyl-n-methylcarbamate, 2-n-methyl-n-boc-amino ethanol, 2-n-boc-n-methylamino ethanol, n-boc-n-methylethanolamine, 2-hydroxyethyl methylcarbamic acid 1,1-dimethylethyl ester, carbamic acid, n-2-hydroxyethyl-n-methyl-, 1,1-dimethylethyl ester
545700
tert-butyl N-(2-hydroxyethyl)-N-methylcarbamate
CC(C)(C)OC(=O)N(C)CCO

N-Boc-N-methylethanolamine is an substituent in the synthesis of anticancer agent, an orally bioavailable small molecule inhibitor of signal transducer and activator of transcription.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-N-methylethanolamine is an substituent in the synthesis of anticancer agent, an orally bioavailable small molecule inhibitor of signal transducer and activator of transcription.

Solubility
Soluble in diochloromethane. Slightly soluble in water.

Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Beilstein 3537964
Solubility Information Soluble in diochloromethane. Slightly soluble in water.
Formula Weight 175.23
Percent Purity 95%
Quantity 1 g
Physical Form Liquid
Chemical Name or Material N-Boc-N-methylethanolamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'