Läs mer
N-Boc-N-methylethylenediamine, 97+%, Thermo Scientific Chemicals
Produktkod. 15442938
Change view
Kvantitet:
1 mL
5 mL
Förpackningsstorlek:
1ml
5ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
15442938 1 mL 1ml
15432938 5 mL 5ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 15442938 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H56456.XF
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 121492-06-6
Molekylformel C8H18N2O2
Molekylvikt (g/mol) 174.244
MDL-nummer MFCD01317789
InChI-nyckel QYJVBVKFXDHFPQ-UHFFFAOYSA-N
Synonym n-boc-n-methylethylenediamine, n-2-aminoethyl-n-methyl carbamic acid tert-butyl ester, n-boc-2-aminoethyl-n-methylamine, tert-butyl n-2-aminoethyl-n-methylcarbamate, n-boc-n-methyl-ethylenediamine, tert-butyl 2-aminoethyl methyl carbamate, 2-n-boc-n-methylamino ethylamine, n-2-aminoethyl-n-methylcarbamicacidtert-butylester, n-boc-n-methyl ethylenediamine
PubChem CID 2756424
IUPAC-namn tert-butyl N-(2-aminoethyl)-N-methylcarbamate
LEDER CC(C)(C)OC(=O)N(C)CCN
Specifikationer
Kemiskt namn eller material N-Boc-N-methylethylenediamine
Brytningsindex 1.447
UN-nummer UN2735
Beilstein 3588326
Densitet 0.975
Kvantitet 1 mL
Flampunkt >100°C (212°F)
Formel vikt 174.24
Procent renhet ≥97%