missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-propargylamine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
N-Boc-propargylamine is used to prepare triazolobenzylidene-thiazolopyrimidines. which act as CDC25 phosphatase inhibitors. Further, it is used for the synthesis of beta-glucan polysaccharide analogs. In addition to this, it is involved in the Pauson-Khand (PK) reaction of norbornadiene and N-Boc-propargylamine to prepare 4,5-disubstituted cyclopentenones .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Boc-propargylamine is used to prepare triazolobenzylidene-thiazolopyrimidines. which act as CDC25 phosphatase inhibitors. Further, it is used for the synthesis of beta-glucan polysaccharide analogs. In addition to this, it is involved in the Pauson-Khand (PK) reaction of norbornadiene and N-Boc-propargylamine to prepare 4,5-disubstituted cyclopentenones .
Solubility
Soluble in chloroform.
Notes
Moisture sensitive. Incompatible with oxidizing agents. Store in a cool place.
N-Boc-propargylamine is used to prepare triazolobenzylidene-thiazolopyrimidines. which act as CDC25 phosphatase inhibitors. Further, it is used for the synthesis of beta-glucan polysaccharide analogs. In addition to this, it is involved in the Pauson-Khand (PK) reaction of norbornadiene and N-Boc-propargylamine to prepare 4,5-disubstituted cyclopentenones .
Solubility
Soluble in chloroform.
Notes
Moisture sensitive. Incompatible with oxidizing agents. Store in a cool place.
Spécification
Melting Point | 40°C to 44°C |
Linear Formula | HC≡CCH2NHCO2C(CH3)3 |
Solubility Information | Soluble in chloroform. |
Formula Weight | 155.2 |
Percent Purity | 97% |
Quantity | 1 g |
Flash Point | 93°C (199°F) |
Sensitivity | Moisture sensitive |
Chemical Name or Material | N-Boc-propargylamine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'