missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-trans-4-(p-toluenesulfonyloxy)-L-proline methyl ester, 98%

Brand:  Alfa Aesar™ H63106.03

Additional Details : CAS Number : 88043-21-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15458126

  • 2313.96 SEK / 1g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

N-Boc-trans-4-(p-toluenesulfonyloxy)-L-proline methyl ester
399.46
7231334
88043-21-4
MFCD02179383
(4R)-4-TsO-N-Boc-Pro-OMe; N-Boc-trans-4-(p-tosyloxy)-L-proline methyl ester

Specifications

N-Boc-trans-4-(p-toluenesulfonyloxy)-L-proline methyl ester
Solid
399.46
7231334
1g
88043-21-4
98%
MFCD02179383
(4R)-4-TsO-N-Boc-Pro-OMe; N-Boc-trans-4-(p-tosyloxy)-L-proline methyl ester
Videos
SDS