missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl benzoate, 98%, Thermo Scientific Chemicals

Catalog Number p-7023447
Click to view available options
Quantity:
250 g
1000 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
136-60-7
C11H14O2
178.23
MFCD00009439
XSIFPSYPOVKYCO-UHFFFAOYSA-N
n-butyl benzoate, benzoic acid, butyl ester, anthrapole az, benzoic acid butyl ester, dai cari xbn, benzoic acid n-butyl ester, butylester kyseliny benzoove, butylbenzoate, unii-1tgz0d0o8i, butylester kyseliny benzoove czech
8698
butyl benzoate
CCCCOC(=O)C1=CC=CC=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
136-60-7
C11H14O2
178.23
MFCD00009439
XSIFPSYPOVKYCO-UHFFFAOYSA-N
n-butyl benzoate, benzoic acid, butyl ester, anthrapole az, benzoic acid butyl ester, dai cari xbn, benzoic acid n-butyl ester, butylester kyseliny benzoove, butylbenzoate, unii-1tgz0d0o8i, butylester kyseliny benzoove czech
8698
butyl benzoate
CCCCOC(=O)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -22°C
Density 1.011
Boiling Point 250°C
Flash Point 106°C (222°F)
Refractive Index 1.498
Quantity 250 g
Beilstein 1867073
Merck Index 14,1552
Formula Weight 178.23
Percent Purity 98%
Chemical Name or Material n-Butyl benzoate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'