Läs mer
N-Butyl Nortadalafil, TRC
Produktkod. 30134430
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134430 100 mg
17943505 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134430 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB693585100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 171596-31-9
Molekylformel C25 H25 N3 O4
Molekylvikt (g/mol) 431.48
Synonym Pyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione, 6-(1,3-benzodioxol-5-yl)-2-butyl-2,3,6,7,12,12a-hexahydro-, (6R,12aR)-, Pyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione, 6-(1,3-benzodioxol-5-yl)-2-butyl-2,3,6,7,12,12a-hexahydro-, (6R-trans)-
IUPAC-namn (2R,8R)-2-(1,3-benzodioxol-5-yl)-6-butyl-3,6,17-triazatetracyclo[8.7.0.0^{3,8}.0^{11,16}]heptadeca-1(10),11,13,15-tetraene-4,7-dione
LEDER CCCCN1CC(=O)N2[C@H](Cc3c([nH]c4ccccc34)[C@H]2c5ccc6OCOc6c5)C1=O
Specifikationer
Kemiskt namn eller material N-Butyl Tadalafil
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C25H25N3O4/c1-2-3-10-27-13-22(29)28-19(25(27)30)12-17-16-6-4-5-7-18(16)26-23(17)24(28)15-8-9-20-21(11-15)32-14-31-20/h4-9,11,19,24,26H,2-3,10,12-14H2,1H3/t19-,24-/m1/s1
Formel vikt 431.1845
Procent renhet >95%