Läs mer
N-Butyldeoxymannojirimycin Hydrochloride, TRC
Produktkod. 30337281
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30337281 5 mg
1 options
Produktkod. 30337281 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB6907505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Butyldeoxymannojirimycin, Hydrochloride
CAS 355012-88-3
Molekylformel C10 H21 N O4 . Cl H
Kvantitet 5 mg
Synonym 3,4,5-Piperidinetriol, 1-butyl-2-(hydroxymethyl)-, hydrochloride (1:1), (2R,3R,4R,5R)-, 3,4,5-Piperidinetriol, 1-butyl-2-(hydroxymethyl)-, hydrochloride, (2R,3R,4R,5R)- (9CI), N-Butyldeoxymannojirimycin hydrochloride
InChI formel InChI=1S/C10H21NO4.ClH/c1-2-3-4-11-5-8(13)10(15)9(14)7(11)6-12;/h7-10,12-15H,2-6H2,1H3;1H/t7-,8-,9-,10-;/m1./s1
LEDER Cl.CCCCN1C[C@@H](O)[C@@H](O)[C@H](O)[C@H]1CO
IUPAC-namn (2R,3R,4R,5R)-1-butyl-2-(hydroxymethyl)piperidine-3,4,5-triol;hydrochloride
Molekylvikt (g/mol) 255.74
Formel vikt 255.1237