Läs mer
N-Butyldeoxynojirimycin Hydrochloride, TRC
Produktkod. 30005771
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30005771 25 mg
30007403 50 mg
17949934 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30005771 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB69100025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 210110-90-0
Molekylformel C10 H21 N O4 . Cl H
Molekylvikt (g/mol) 255.74
Synonym 3,4,5-Piperidinetriol, 1-butyl-2-(hydroxymethyl)-, hydrochloride (1:1), (2R,3R,4R,5S)-, 3,4,5-Piperidinetriol, 1-butyl-2-(hydroxymethyl)-, hydrochloride, (2R,3R,4R,5S)- (9CI), Miglustat hydrochloride
IUPAC-namn (2R,3R,4R,5S)-1-butyl-2-(hydroxymethyl)piperidine-3,4,5-triol;hydrochloride
LEDER Cl.CCCCN1C[C@H](O)[C@@H](O)[C@H](O)[C@H]1CO
Specifikationer
Kemiskt namn eller material N-Butyldeoxynojirimycin Hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C10H21NO4.ClH/c1-2-3-4-11-5-8(13)10(15)9(14)7(11)6-12;/h7-10,12-15H,2-6H2,1H3;1H/t7-,8+,9-,10-;/m1./s1
Formel vikt 255.1237
Procent renhet >95%