missing translation for 'onlineSavingsMsg'
Learn More

n-Butyldimethylchlorosilane, 96%, Thermo Scientific Chemicals

Catalog Number p-7027079
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
1000-50-6
C6H15ClSi
150.72
MFCD00015727
MXOSTENCGSDMRE-UHFFFAOYSA-N
butylchlorodimethylsilane, butyldimethylchlorosilane, n-butyldimethylchlorosilane, butyldimethylsilyl chloride, 1-butyldimethylchlorosilane, silane, butylchlorodimethyl, butyl chloro dimethylsilane, n-butyl-dimethylchlorosilane, butyl-chloro-dimethyl-silane, n-butyldimethylsilyl chloride
66079
CCCC[Si](C)(C)Cl
This item is not returnable. View return policy
1000-50-6
C6H15ClSi
150.72
MFCD00015727
MXOSTENCGSDMRE-UHFFFAOYSA-N
butylchlorodimethylsilane, butyldimethylchlorosilane, n-butyldimethylchlorosilane, butyldimethylsilyl chloride, 1-butyldimethylchlorosilane, silane, butylchlorodimethyl, butyl chloro dimethylsilane, n-butyl-dimethylchlorosilane, butyl-chloro-dimethyl-silane, n-butyldimethylsilyl chloride
66079
CCCC[Si](C)(C)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 138°C
Quantity 5 g
UN Number UN2920
Beilstein 1732573
Flash Point 27°C (80°F)
IUPAC Name butyl(chloro)dimethylsilane
Formula Weight 150.73
Percent Purity 96%
Refractive Index 1.42
Sensitivity Moisture sensitive
Density 0.875
Chemical Name or Material n-Butyldimethylchlorosilane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'