missing translation for 'onlineSavingsMsg'
Learn More

N-Butylimidazole, 98%, Thermo Scientific Chemicals

Catalog Number p-7077286
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
4316-42-1
C7H12N2
124.19
MFCD00042753
MCMFEZDRQOJKMN-UHFFFAOYSA-N
1-butyl-1h-imidazole, 1h-imidazole, 1-butyl, n-butylimidazole, 1-n-butylimidazole, n-n-butyl imidazole, 1-n-butyl imidazole, 1-buthylimidazole, n-n-butylimidazole, imidazole, 1-butyl
61347
1-butyl-1H-imidazole
CCCCN1C=CN=C1
This item is not returnable. View return policy
4316-42-1
C7H12N2
124.19
MFCD00042753
MCMFEZDRQOJKMN-UHFFFAOYSA-N
1-butyl-1h-imidazole, 1h-imidazole, 1-butyl, n-butylimidazole, 1-n-butylimidazole, n-n-butyl imidazole, 1-n-butyl imidazole, 1-buthylimidazole, n-n-butylimidazole, imidazole, 1-butyl
61347
1-butyl-1H-imidazole
CCCCN1C=CN=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.9450g/mL
Boiling Point 257.0°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 25 g
Specific Gravity 0.945
Solubility Information Solubility in water: soluble.
Formula Weight 124.19
Percent Purity 98%
Chemical Name or Material N-Butylimidazole
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N-Butylimidazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'