Läs mer
N-Butyroyl Phytosphingosine, TRC
Produktkod. 30179564
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30179564 5 mg
1 options
Produktkod. 30179564 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB8025155MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 409085-57-0
Molekylformel C22H45NO4
Molekylvikt (g/mol) 387.6
Synonym N-[(1S,2S,3R)-2,3-Dihydroxy-1-(hydroxymethyl)heptadecyl]butanamide, C4-Phytoceramide;
LEDER OC[C@H](NC(CCC)=O)[C@H](O)[C@H](O)CCCCCCCCCCCCCC
Specifikationer
Kemiskt namn eller material N-Butyroyl Phytosphingosine