Läs mer
Invitrogen™ N-cadherin Monoclonal Antibody (8C11)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA12002
Beskrivning
MA1-2002 detects N-Cadherin from human neuronal samples, including SHSY5Y cells. MA1-2002 also detects N-Cadherin in rabbit and hamster brain lysates. MA1-2002 does not detect N-Cadherin in lysates from mouse, rat, pig and cow brain. This antibody is not suitable for mouse C2C12 cell lysates in Western blot analysis. MA1-2002 has been successfully used in Western blot, Immunoprecipitation, IHC (P) and Immunofluorescence applications. By Western blot, this antibody detects N-Cadherin at 130 kDa. This antibody is not suitable for frozen tissues in immunohistochemistry applications. The MA1-2002 immunogen is a bacterial fusion protein to the extracellular domain of N-cadherin (a domain between EC3 and EC4, amino acid residues 92-593 from human N-cadherin).
N-cadherin is a 140 kDa protein belonging to a family of transmembrane molecules that mediate calcium-dependent intercellular adhesion. Cadherins are involved in controlling morphogenetic movements during development and regulate cell surface adhesion through homotypic adhesion with the same cadherin species. N-cadherin's function is dependent on its association with the actin-cytoskeleton and is mediated through interactions between the C-terminal region of N-cadherin and the cytoplasmic catenin proteins. The stability of this association is regulated by phosphorylation and dephosphorylation of beta-catenin. Further, N-cadherin is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. The protein functions during gastrulation and is required for establishment of left-right asymmetry.
Specifikationer
| N-cadherin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1mg/mL BSA and 0.05% sodium azide | |
| O55075, P15116, P19022 | |
| CDH2 | |
| Bacterial fusion protein corresponding to the extracellular domain of N-cadherin. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rabbit, Hamster | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 8C11 | |
| Unconjugated | |
| CDH2 | |
| bib; cadherin 2; cadherin 2 L homeolog; cadherin 2 type 1 N-cadherin (neuronal); cadherin 2, neuronal; cadherin 2, type 1 preproprotein; cadherin 2, type 1, N-cadherin (neuronal); cadherin 2, type 1, N-cadherin (neuronal) L homeolog; cadherin 2, type 1, N-cadherin a (neuronal); Cadherin2; cadherin-2; Cadherin-2A; Cadherin-N; calcium-dependent adhesion protein, neuronal; CD325; CDH2; cdh2.L; cdh2a; cdh2-a; cdh2-b; CDHN; cdhn-a; CDw325; CDw325 antigen; cell adhesion molecule; glass onion; glo; I79_014914; labyrinth; lyr; NCAD; N-Cad; N-cadherin; N-cadherin 1; N-cadherin A; N-cadherin precursor; N-cadherin-2; neural cadherin; Neural cadherin A; neural cadherin-2; neural calcium-dependent cell adhesion molecule; neural-cadherin; pac; Parachute; unnamed protein product; wu:fb47h04; XELAEV_18031953mg; ZNCAD | |
| Mouse | |
| Protein G | |
| RUO | |
| 1000, 100344571, 100689204, 12558 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.