Läs mer
N-cadherin Mouse anti-Human, CoraLite 488, Clone: 1D8B3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66219-100UL
Beskrivning
The N-cadherin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human N-cadherin. This antibody recognizes human, pig, mouse, rat antigen. The N-cadherin antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifikationer
| N-cadherin | |
| Monoclonal | |
| CoraLite 488 | |
| 100 μL | |
| 1000 | |
| IgG1 |
| Immunofluorescence | |
| 1D8B3 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.