Läs mer
N-Carbamoyl Carbamazepine, TRC
Produktkod. 30355928
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30355928 5 mg
1 options
Produktkod. 30355928 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC1758605MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (5H-Dibenzo[b,f]azepin-5-yl-carbonyl)urea (N-Carbamoylcarbamazepine)
CAS 1219170-51-0
Molekylformel C16 H13 N3 O2
Kvantitet 5 mg
Synonym (5H-Dibenzo[b,f]azepin-5-yl-carbonyl)urea, Carbamazepine Imp. C (EP), N-Carbamoylcarbamazepine
InChI formel InChI=1S/C16H13N3O2/c17-15(20)18-16(21)19-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)19/h1-10H,(H3,17,18,20,21)
LEDER NC(=O)NC(=O)N1c2ccccc2C=Cc3ccccc13
IUPAC-namn N-carbamoylbenzo[b][1]benzazepine-11-carboxamide
Molekylvikt (g/mol) 279.29
Formel vikt 279.1008