Läs mer
N-Carbamoyl-L-cysteine, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-Carbamoyl-L-cysteine |
| CAS | 24583-23-1 |
| Molekylformel | C4 H8 N2 O3 S |
| Kvantitet | 500 mg |
| Synonym | L-Cysteine, N-(aminocarbonyl)-, Cysteine, N-carbamoyl-, L- (8CI), N-(Aminocarbonyl)-L-cysteine, L-Cysteine-N-carbamide, L-α-Ureido-β-mercaptopropionic acid, N-(Aminocarbonyl)cysteine, N-Carbamoyl-L-cysteine |
| InChI formel | InChI=1S/C4H8N2O3S/c5-4(9)6-2(1-10)3(7)8/h2,10H,1H2,(H,7,8)(H3,5,6,9)/t2-/m0/s1 |
| LEDER | NC(=O)N[C@@H](CS)C(=O)O |
| IUPAC-namn | (2R)-2-(carbamoylamino)-3-sulfanylpropanoic acid |
| Molekylvikt (g/mol) | 164.18 |
| Formel vikt | 164.0256 |