Läs mer
N-Carbamoyl Linagliptin, TRC
Produktkod. 30368689
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30368689 50 mg
30352603 100 mg
30355157 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30368689 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC17609050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1955514-80-3
Molekylformel C26H29N9O3
Molekylvikt (g/mol) 515.57
Synonym Linagliptin N-Aminocarbonyl Impurity, N-[1-[7-(2-Butyn-1-yl)-2,3,6,7-tetrahydro-3-methyl-1-[(4-methyl-2-quinazolinyl)methyl]-2,6-dioxo-1H-purin-8-yl]-3-piperidinyl]urea
IUPAC-namn [1-[7-but-2-ynyl-3-methyl-1-[(4-methylquinazolin-2-yl)methyl]-2,6-dioxopurin-8-yl]piperidin-3-yl]urea
LEDER CC#CCn1c(nc2N(C)C(=O)N(Cc3nc(C)c4ccccc4n3)C(=O)c12)N5CCC[C@H](C5)NC(=O)N
Specifikationer
Kemiskt namn eller material N-Carbamoyl Linagliptin
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C26H29N9O3/c1-4-5-13-34-21-22(31-25(34)33-12-8-9-17(14-33)29-24(27)37)32(3)26(38)35(23(21)36)15-20-28-16(2)18-10-6-7-11-19(18)30-20/h6-7,10-11,17H,8-9,12-15H2,1-3H3,(H3,27,29,37)
Formel vikt 515.2393
Procent renhet >95%