Läs mer
N-Carbobenzoxy-dl-leucine, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-Carbobenzoxy-dl-leucine |
| CAS | 3588-60-1 |
| Molekylformel | C14H19NO4 |
| Synonym | DL-Leucine, N-[(phenylmethoxy)carbonyl]-, Leucine, N-carboxy-, N-benzyl ester, DL- (8CI), N-[(Phenylmethoxy)carbonyl]leucine, DL-(Carbobenzyloxy)leucine, DL-Cbz leucine, DL-N-(Benzyloxycarbonyl)leucine, N-(Benzyloxycarbonyl)-DL-leucine |
| InChI formel | InChI=1S/C14H19NO4/c1-10(2)8-12(13(16)17)15-14(18)19-9-11-6-4-3-5-7-11/h3-7,10,12H,8-9H2,1-2H3,(H,15,18)(H,16,17) |
| LEDER | CC(C)CC(NC(=O)OCc1ccccc1)C(=O)O |
| IUPAC-namn | 4-methyl-2-(phenylmethoxycarbonylamino)pentanoic acid |
| Molekylvikt (g/mol) | 265.3 |
| Kvantitet | 500 mg |
| Formel vikt | 265.1314 |
| Visa mer |