Läs mer
N-Carbobenzoxy-L-tryptophanamide, TRC
Produktkod. 30367612
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30367612 100 mg
1 options
Produktkod. 30367612 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC176003100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Carbobenzoxy-L-tryptophanamide
CAS 20696-64-4
Molekylformel C19H19N3O3
Kvantitet 100 mg
Synonym L-(1-carbamoyl-2-indol-3-ylethyl)carbamic Acid Benzyl Ester, [(2S)-2-amino-1-(1H-indol-3-ylmethyl)-2-oxoethyl]carbamic Acid Phenylmethyl Ester, N-(Benzyloxycarbonyl)tryptophanamide
InChI formel InChI=1S/C19H19N3O3/c20-18(23)17(10-14-11-21-16-9-5-4-8-15(14)16)22-19(24)25-12-13-6-2-1-3-7-13/h1-9,11,17,21H,10,12H2,(H2,20,23)(H,22,24)/t17-/m0/s1
LEDER NC(=O)[C@H](Cc1c[nH]c2ccccc12)NC(=O)OCc3ccccc3
IUPAC-namn benzyl N-[(2S)-1-amino-3-(1H-indol-3-yl)-1-oxopropan-2-yl]carbamate
Molekylvikt (g/mol) 337.37
Formel vikt 337.1426