Läs mer
N-Carboxybenzyl D-Valacyclovir, TRC
Produktkod. 30349565
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30349565 10 mg
1 options
Produktkod. 30349565 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC17799510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Carboxybenzyl D-Valacyclovir
CAS 124832-32-2
Molekylformel C21 H26 N6 O6
Kvantitet 10 mg
Synonym N(Val)-Carboxybenzyl Valacyclovir, N-[(Phenylmethoxy)carbonyl]-D-valine 2-[(2-Amino-1,6-dihydro-6-oxo-9H-purin-9-yl)methoxy]Ethyl Ester, Valacyclovir Related Compound E
InChI formel InChI=1S/C21H26N6O6/c1-13(2)15(24-21(30)33-10-14-6-4-3-5-7-14)19(29)32-9-8-31-12-27-11-23-16-17(27)25-20(22)26-18(16)28/h3-7,11,13,15H,8-10,12H2,1-2H3,(H,24,30)(H3,22,25,26,28)/t15-/m1/s1
LEDER CC(C)[C@@H](NC(=O)OCc1ccccc1)C(=O)OCCOCn2cnc3c(O)nc(N)nc23
IUPAC-namn 2-[(2-amino-6-hydroxypurin-9-yl)methoxy]ethyl (2R)-3-methyl-2-(phenylmethoxycarbonylamino)butanoate
Molekylvikt (g/mol) 458.468
Formel vikt 458.191