Läs mer
N-Cbz-D-isoleucine, TRC
Produktkod. 30179527
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179527 50 mg
30178963 500 mg
30179611 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179527 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC22755550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 28862-89-7
Molekylformel C14 H19 N O4
Molekylvikt (g/mol) 265.3
Synonym N-[(Phenylmethoxy)carbonyl]-D-isoleucine, N-Carboxy-D-isoleucine N-Benzyl Ester, CBZ-D-Isoleucine, Cbz-D-Ile-OH, N-Benzyloxycarbonyl-D-isoleucine, N-Carbobenzoxy-D-isoleucine, N-Carbobenzyloxy-D-isoleucine
IUPAC-namn (2R,3R)-3-methyl-2-(phenylmethoxycarbonylamino)pentanoic acid
LEDER CC[C@@H](C)[C@@H](NC(=O)OCc1ccccc1)C(=O)O
Specifikationer
Kemiskt namn eller material N-Cbz-D-isoleucine
InChI formel InChI=1S/C14H19NO4/c1-3-10(2)12(13(16)17)15-14(18)19-9-11-7-5-4-6-8-11/h4-8,10,12H,3,9H2,1-2H3,(H,15,18)(H,16,17)/t10-,12-/m1/s1
Kvantitet 50 mg
Formel vikt 265.1314