Learn More
N-Cbz-L-leucine, TRC
Product Code. 30362950
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity Unit Size
30362950 10 g 10g
1 options
Product Code. 30362950 Supplier Toronto Research Chemicals Supplier No. TRCC22776010G
 Restricted Item
Beräknad leverans: 09-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Benzyloxycarbonyl-L-leucine
CAS 2018-66-8
Molecular Formula C14 H19 N O4
Quantity 10 g
Synonym N-Benzyloxycarbonyl-L-leucine, L-Leucine, N-[(phenylmethoxy)carbonyl]-, Leucine, N-carboxy-, N-benzyl ester (6CI), Leucine, N-carboxy-, N-benzyl ester, L- (7CI,8CI), N-[(Phenylmethoxy)carbonyl]-L-leucine, (Benzyloxycarbonyl)leucine, (Carbobenzoxy)leucine, (Carbobenzyloxy)-L-leucine, Benzyloxycarbonyl-L-leucine, CBZ-L-leucine, Carbobenzoxy-L-leucine, L-(Carbobenzyloxy)leucine, L-N-(Benzyloxycarbonyl)leucine, N-(Benzyloxycarbonyl)-L-leucine, N-(Benzyloxycarbonyl)leucine, N-(Carbobenzoxy)leucine, N-Carbobenzoxy-L-leucine, N-Carbobenzyloxy-L-leucine, N-Carboxy-L-leucine N-benzyl ester, NSC 60039, CBZ-L-leucine
InChI Formula InChI=1S/C14H19NO4/c1-10(2)8-12(13(16)17)15-14(18)19-9-11-6-4-3-5-7-11/h3-7,10,12H,8-9H2,1-2H3,(H,15,18)(H,16,17)/t12-/m0/s1
SMILES CC(C)C[C@H](NC(=O)OCc1ccccc1)C(=O)O
IUPAC Name (2S)-4-methyl-2-(phenylmethoxycarbonylamino)pentanoic acid
Molecular Weight (g/mol) 265.3
Formula Weight 265.1314