Läs mer
N-Cbz-L-leucin, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-Benzyloxycarbonyl-L-leucine |
| CAS | 2018-66-8 |
| Molekylformel | C14 H19 N O4 |
| Kvantitet | 25 g |
| Synonym | N-Benzyloxycarbonyl-L-leucine, L-Leucine, N-[(phenylmethoxy)carbonyl]-, Leucine, N-carboxy-, N-benzyl ester (6CI), Leucine, N-carboxy-, N-benzyl ester, L- (7CI,8CI), N-[(Phenylmethoxy)carbonyl]-L-leucine, (Benzyloxycarbonyl)leucine, (Carbobenzoxy)leucine, (Carbobenzyloxy)-L-leucine, Benzyloxycarbonyl-L-leucine, CBZ-L-leucine, Carbobenzoxy-L-leucine, L-(Carbobenzyloxy)leucine, L-N-(Benzyloxycarbonyl)leucine, N-(Benzyloxycarbonyl)-L-leucine, N-(Benzyloxycarbonyl)leucine, N-(Carbobenzoxy)leucine, N-Carbobenzoxy-L-leucine, N-Carbobenzyloxy-L-leucine, N-Carboxy-L-leucine N-benzyl ester, NSC 60039, CBZ-L-leucine |
| InChI formel | InChI=1S/C14H19NO4/c1-10(2)8-12(13(16)17)15-14(18)19-9-11-6-4-3-5-7-11/h3-7,10,12H,8-9H2,1-2H3,(H,15,18)(H,16,17)/t12-/m0/s1 |
| LEDER | CC(C)C[C@H](NC(=O)OCc1ccccc1)C(=O)O |
| IUPAC-namn | (2S)-4-metyl-2-(fenylmetoxykarbonylamino)pentansyra |
| Molekylvikt (g/mol) | 265.3 |
| Formel vikt | 265.1314 |