Läs mer
N-Cbz-N'-isopropyl-DL-phenylglycinamide, TRC
Produktkod. 30351595
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30351595 100 mg
1 options
Produktkod. 30351595 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC227590100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Cbz-N'-isopropyl-DL-phenylglycinamide
CAS 1393441-67-2
Molekylformel C19H22N2O3
Kvantitet 100 mg
Synonym Benzyl 2-(Isopropylamino)-2-oxo-1-phenylethylcarbamate
InChI formel InChI=1S/C19H22N2O3/c1-14(2)20-18(22)17(16-11-7-4-8-12-16)21-19(23)24-13-15-9-5-3-6-10-15/h3-12,14,17H,13H2,1-2H3,(H,20,22)(H,21,23)
LEDER CC(C)NC(=O)C(NC(=O)OCc1ccccc1)c2ccccc2
IUPAC-namn benzyl N-[2-oxo-1-phenyl-2-(propan-2-ylamino)ethyl]carbamate
Molekylvikt (g/mol) 326.39
Formel vikt 326.163