Läs mer
N-Chloro Ethyl ThioTEPA, TRC
Produktkod. 30136169
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136169 25 mg
30137040 5 mg
30004189 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136169 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC36417025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 90877-51-3
Molekylformel C6H13ClN3PS
Molekylvikt (g/mol) 225.68
Synonym ThioTEPA Chloroethyl Analog, ThioTEPA Chloro Impurity, P,P-Bis(1-aziridinyl)-N-(2-chloroethyl)-phosphinothioic Amide, N,N’-Diethylene-N’’-2-chloroethyl Thiophosphoramide, N-(2-Chloroethyl)-P,P-bisaziridinyl Thiophosphamide
IUPAC-namn N-[bis(aziridin-1-yl)phosphinothioyl]-2-chloroethanamine
LEDER S=[P](N1CC1)(N2CC2)NCCCl
Specifikationer
Kemiskt namn eller material N-Chloro Ethyl ThioTEPA
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C6H13ClN3PS/c7-1-2-8-11(12,9-3-4-9)10-5-6-10/h1-6H2,(H,8,12)
Formel vikt 225.0256
Procent renhet >95%