Läs mer
N-(Chloroacetoxy)succinimide, TRC
Produktkod. 30359039
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30359039 1 g
1 options
Produktkod. 30359039 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC3633751G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-(Chloroacetoxy)succinimide
CAS 27243-15-8
Molekylformel C6 H6 Cl N O4
Kvantitet 1 g
Synonym 2-Chloroacetic Acid 2,5-Dioxo-1-pyrrolidinyl Ester, 2-Chloroacetic Acid N-Hydroxysuccinimide Ester, 1-[(Chloroacetyl)oxy]-2,5-pyrrolidinedione, N-Succinimidyl 2-chloroacetate
InChI formel InChI=1S/C6H6ClNO4/c7-3-6(11)12-8-4(9)1-2-5(8)10/h1-3H2
LEDER ClCC(=O)ON1C(=O)CCC1=O
IUPAC-namn (2,5-dioxopyrrolidin-1-yl) 2-chloroacetate
Molekylvikt (g/mol) 191.57
Formel vikt 190.9985