Läs mer
N-Chloroacetyl-tert-butylamine, TRC
Produktkod. 30368360
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30368360 100 mg
1 options
Produktkod. 30368360 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC363390100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Chloroacetyl-tert-butylamine
CAS 15678-99-6
Molekylformel C6H12ClNO
Kvantitet 100 mg
Synonym alpha-Chloro-N-tert-butylacetamide, 2-Chloro-N-t-butylacetamide, N-Chloroacetyl-tert-butylamine, N-tert-Butyl-2-chloroacetamide, NSC 8361
InChI formel InChI=1S/C6H12ClNO/c1-6(2,3)8-5(9)4-7/h4H2,1-3H3,(H,8,9)
LEDER CC(C)(C)NC(=O)CCl
IUPAC-namn N-tert-butyl-2-chloroacetamide
Molekylvikt (g/mol) 149.06
Formel vikt 149.0607