Läs mer
N-Chlorobetainyl chloride, TRC
Produktkod. 30360234
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30360234 1 g
1 options
Produktkod. 30360234 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC6518031G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Chlorobetainyl chloride
CAS 53684-57-4
Molekylformel C5 H11 Cl N O . Cl
Kvantitet 1 g
Synonym 2-Chloro-N,N,N-trimethyl-2-oxo-ethanaminium Chloride, [(Chloroformyl)methyl]trimethylammonium Chloride
InChI formel InChI=1S/C5H11ClNO.ClH/c1-7(2,3)4-5(6)8;/h4H2,1-3H3;1H/q+1;/p-1
LEDER [Cl-].C[N+](C)(C)CC(=O)Cl
IUPAC-namn (2-chloro-2-oxoethyl)-trimethylazanium;chloride
Molekylvikt (g/mol) 172.053
Formel vikt 171.022