Läs mer
N-Chloromethyl Hydroxychloroquine, TRC
Produktkod. 30009764
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009764 10 mg
30005907 50 mg
30004744 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009764 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC60220610MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1159208-89-5
Molekylformel C19H28Cl2IN3O
Molekylvikt (g/mol) 512.26
IUPAC-namn chloromethyl-[4-[(7-chloroquinolin-4-yl)amino]pentyl]-ethyl-(2-hydroxyethyl)azanium;iodide
LEDER CC[N+](CCl)(CCCC(C)NC1=C2C=CC(Cl)=CC2=NC=C1)CCO.[I-]
Specifikationer
Kemiskt namn eller material N-Chloromethyl Hydroxychloroquine Iodide
Kvantitet 10 mg
InChI formel InChI=1S/C19H28Cl2N3O.HI/c1-3-24(14-20,11-12-25)10-4-5-15(2)23-18-8-9-22-19-13-16(21)6-7-17(18)19;/h6-9,13,15,25H,3-5,10-12,14H2,1-2H3,(H,22,23);1H/q+1;/p-1