Läs mer
N-Cyano-3-(cyanoamino)-N'-methyl-7,8-dithia-2,4,11-triazadodec-2-en-12-imidamide (Cimetidine Impurity), TRC
Produktkod. 30352806
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30352806 25 mg
30359934 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30352806 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC98827525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 74886-59-2
Molekylformel C10 H18 N8 S2
Molekylvikt (g/mol) 314.43
Synonym 1,1'-(Disulphanediyldiethylene)bis(2-cyano-3-methylguanidine), Cimetidine Hydrochloride Imp. H (EP)
IUPAC-namn 2-cyano-1-[2-[2-[(N'-cyano-N-methylcarbamimidoyl)amino]ethyldisulfanyl]ethyl]-3-methylguanidine
LEDER CNC(=NC#N)NCCSSCCNC(=NC#N)NC
Specifikationer
Kemiskt namn eller material 1,1'-(Disulfanediyldiethylene)bis(2-cyano-3-methylguanidine)
Kvantitet 25 mg
InChI formel InChI=1S/C10H18N8S2/c1-13-9(17-7-11)15-3-5-19-20-6-4-16-10(14-2)18-8-12/h3-6H2,1-2H3,(H2,13,15,17)(H2,14,16,18)
Formel vikt 314.1096