missing translation for 'onlineSavingsMsg'
Läs mer

N-Cyanonor-LSD, TRC

Produktkod. 30358290
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
2.5 mg
Förpackningsstorlek:
2.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30358290 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30358290 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC9820982.5MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material N-Cyanonor-LSD
CAS 35779-41-0
Molekylformel C20H22N4O
Kvantitet 2.5 mg
Synonym N6-Cyano-N6-demethyl-LSD, (8beta)-6-Cyano-9,10-didehydro-N,N-diethyl-ergoline-8-carboxamide, Indolo[4,3-fg]quinoline Ergoline-8-carboxamide Derivative, (6aR,9R)-7-Cyano-N,N-diethyl-4,6,6a,7,8,9-hexahydroindolo[4,3-fg]quinoline-9-carboxamide
InChI formel InChI=1S/C20H22N4O/c1-3-23(4-2)20(25)14-8-16-15-6-5-7-17-19(15)13(10-22-17)9-18(16)24(11-14)12-21/h5-8,10,14,18,22H,3-4,9,11H2,1-2H3/t14-,18-/m1/s1
LEDER CCN(CC)C(=O)[C@H]1CN(C#N)[C@@H]2Cc3c[nH]c4cccc(C2=C1)c34
IUPAC-namn (6aR,9R)-7-cyano-N,N-diethyl-6,6a,8,9-tetrahydro-4H-indolo[4,3-fg]quinoline-9-carboxamide
Molekylvikt (g/mol) 334.41
Formel vikt 334.1794
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.