missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Cycloheptyl-2-nitrobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H60568.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15497048

  • 6855.28 SEK / 1g


Estimated Shipment date
from Supplier 25-04-2019
Add to basket

Specifications

Chemical Identifiers

346691-67-6
298.36
WAUMMPTYQUZBCV-UHFFFAOYSA-N
767756
C13H18N2O4S
MFCD00784355
n-cycloheptyl-2-nitrobenzenesulfonamide, cambridge id 5350324, n-cycloheptyl-2-nitro-benzenesulfonamide, cycloheptyl 2-nitrophenyl sulfonyl amine

Specifications

N-Cycloheptyl-2-nitrobenzenesulfonamide
346691-67-6
MFCD00784355
WAUMMPTYQUZBCV-UHFFFAOYSA-N
767756
Solid
1g
C13H18N2O4S
n-cycloheptyl-2-nitrobenzenesulfonamide, cambridge id 5350324, n-cycloheptyl-2-nitro-benzenesulfonamide, cycloheptyl 2-nitrophenyl sulfonyl amine
298.36
298.36
97%
Videos
SDS