Läs mer
N-Cycloheptylmethylamine Hydrochloride, TRC
Produktkod. 30475666
Change view
Kvantitet:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30475666 1 g
30467416 10 g
30474914 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30475666 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM1032741G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1255718-16-1
Molekylformel C8H17N . HCl
Molekylvikt (g/mol) 163.69
Synonym N-methyl-cycloheptanamine Hydrochloride
IUPAC-namn N-methylcycloheptanamine;hydrochloride
LEDER Cl.CNC1CCCCCC1
Specifikationer
Kemiskt namn eller material N-Cycloheptylmethylamine Hydrochloride
Kvantitet 1 g
InChI formel InChI=1S/C8H17N.ClH/c1-9-8-6-4-2-3-5-7-8;/h8-9H,2-7H2,1H3;1H
Formel vikt 163.1128