missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Cyclohexyl-2,5-difluorobenzamide, 97%

Brand:  Alfa Aesar™ H61029.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15457518

  • 6228.40 SEK / 1g


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Specifications

Chemical Identifiers

N-Cyclohexyl-2,5-difluorobenzamide
239.27
MFCD11842148

Specifications

N-Cyclohexyl-2,5-difluorobenzamide
97%
MFCD11842148
Solid
239.27
1g
Videos
SDS