Läs mer
N-Cyclohexylbenzo[d]thiazole-2-sulfonamide, TRC
Produktkod. 30365469
Change view
Kvantitet:
250 mg
500 mg
2.5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30365469 250 mg -
30351728 500 mg -
30349430 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30365469 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC992305250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 95-33-0
Molekylformel C13 H16 N2 S2
Molekylvikt (g/mol) 264.41
Synonym N-Cyclohexyl-2-benzothiazolylsulfenamide, 2-Benzothiazolesulfenamide, N-cyclohexyl-, N-Cyclohexyl-2-benzothiazolesulfenamide, 2-(Cyclohexylaminothio)benzothiazole, 2-Benzothiazolesulfenic acid N-cyclohexylamide, Accel CZ, Accelerator CBS, Accelerator CM, Accelerator CZ, Accicure HBS, Banac CBS, Benzothiazyl-2-cyclohexylsulfenamide, CBS, CBS (accelerator), CBS 80, CBTS, CZ, CZ 80, CZ Vulcanizer, CZ-G, Conac A, Conac S, Curax, Delac S, Durax, Ekagom CBS, N-Cyclohexyl-2-benzothiazolylsulfenamide, N-Cyclohexyl-2-benzothiazolylsulphenamide, N-Cyclohexyl-2-benzothiazylsulfenamide, N-Cyclohexylbenzothiazole-2-sulphenamide, N-Cyclohexylbenzothiazolesulfenamide, NSC 4809, Nocceler CZ, Nocceler CZ-G, Nocceler CZ-P, Nocceler CZ-R, Oricel CZ, Pennac CBS, Perkacit CBS, Rhenogran CBS, Rhenogran CBS 80, Rhodifax 16, Royal CBTS, Sanceler CM, Sanceler CM-G, Sanceler CM-PO, Sanceler CZ-G, Sanceler CZ-P, Santocure, Santocure CBS, Soxinol CZ, Sulfenamide Ts, Sulfenax, Sulfenax CB, Sulfenax CB 30, Sulfenax CB/K, Thiohexam, Vulcafor CBS, Vulcafor HBS, Vulkacit C, Vulkacit CZ, Vulkacit CZ/C, Vulkacit CZ/CV, Vulkacit CZ/EG, Vulkacit CZ/EG-C, Vulkacit CZ/K, Vulkafil ZN 94TT02
IUPAC-namn N-(1,3-benzothiazol-2-ylsulfanyl)cyclohexanamine
LEDER C1CCC(CC1)NSc2nc3ccccc3s2
Specifikationer
Kemiskt namn eller material N-Cyclohexylbenzo[d]thiazole-2-sulfonamide
Anteckningar om renhetsgrad HPLC
Kvantitet 250 mg
InChI formel InChI=1S/C13H16N2S2/c1-2-6-10(7-3-1)15-17-13-14-11-8-4-5-9-12(11)16-13/h4-5,8-10,15H,1-3,6-7H2
Formel vikt 264.0755
Procent renhet >95%