Läs mer
N-(Cyclohexylthio)phthalimide, TRC
Produktkod. 30470455
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30470455 10 g
1 options
Produktkod. 30470455 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN27283010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-cyclohexylsulfanylisoindole-1,3-dione
CAS 17796-82-6
Molekylformel C14 H15 N O2 S
Kvantitet 10 g
Synonym N-(Cyclohexylthio)phthalimide, 2-(Cyclohexylthio)-1H-isoindole-1,3(2H)-dione, 2-(Cyclohexylsulfanyl)-2,3-dihydro-1H-isoindole-1,3-dione, 2-(Cyclohexylthio)isoindoline-1,3-dione, 2-Cyclohexylsulfanylisoindole-1,3-dione, Accitard RE, Anscorch CTP, CTP, CTP (chinese inhibitor), CTP-PVI, Duslin P, Duslin PP, N-(Cyclohexylthio)phthalimide, N-Cyclohexylsulfenylphthalimide, PVI, Pilgard PVI, Retarder CTP, Rhenogran CPT 80, Santogard PVI, Santogard PVI-DS, Vulkalent G, Vulkalent P
InChI formel InChI=1S/C14H15NO2S/c16-13-11-8-4-5-9-12(11)14(17)15(13)18-10-6-2-1-3-7-10/h4-5,8-10H,1-3,6-7H2
LEDER O=C1N(SC2CCCCC2)C(=O)c3ccccc13
IUPAC-namn 2-cyclohexylsulfanylisoindole-1,3-dione
Molekylvikt (g/mol) 261.339
Formel vikt 261.082