missing translation for 'onlineSavingsMsg'
Learn More

N-Decane, Technical, Fisher Chemical™

Catalog Number p-7072959
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
124-18-5
C10H22
8954
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
124-18-5
C10H22
8954
Melting Point -30°C
Color Colorless
Boiling Point 174°C
Packaging Amber glass bottle
Quantity 100 mL
Formula Weight 142.28g/mol
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • Decane
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H226-Flammable liquid and vapour.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'