Läs mer
N-Decarboxymethylated Indoxacarb, TRC
Produktkod. 30355084
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30355084 100 mg
1 options
Produktkod. 30355084 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD101414100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (S)-Indoxacarb metabolite IN-JT 333
CAS 200568-74-7
Molekylformel C20 H15 Cl F3 N3 O5
Kvantitet 100 mg
Synonym Indeno[1,2-e][1,3,4]oxadiazine-4a(3H)-carboxylic acid, 7-chloro-2,5-dihydro-2-[[[4-(trifluoromethoxy)phenyl]amino]carbonyl]-, methyl ester, (S)-, DCJW, N-Decarboxymethylated indoxacarb
InChI formel InChI=1S/C20H15ClF3N3O5/c1-30-17(28)19-9-11-8-12(21)2-7-15(11)16(19)26-27(10-31-19)18(29)25-13-3-5-14(6-4-13)32-20(22,23)24/h2-8H,9-10H2,1H3,(H,25,29)/t19-/m0/s1
LEDER COC(=O)[C@]12Cc3cc(Cl)ccc3C1=NN(CO2)C(=O)Nc4ccc(OC(F)(F)F)cc4
IUPAC-namn methyl (4aS)-7-chloro-2-[[4-(trifluoromethoxy)phenyl]carbamoyl]-3,5-dihydroindeno[1,2-e][1,3,4]oxadiazine-4a-carboxylate
Molekylvikt (g/mol) 469.8
Formel vikt 469.0652