Läs mer
N-Deethoxymethyl Chlorfenapyr, TRC
Produktkod. 30356662
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30356662 50 mg
30354515 25 mg
30354930 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30356662 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD22849850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 122454-29-9
Molekylformel C12 H5 Br Cl F3 N2
Molekylvikt (g/mol) 349.53
Synonym 1H-Pyrrole-3-carbonitrile, 4-bromo-2-(4-chlorophenyl)-5-(trifluoromethyl)-, 4-Bromo-2-(4-chlorophenyl)-5-(trifluoromethyl)-1H-pyrrole-3-carbonitrile, 4-Bromo-2-(4-chlorophenyl)-3-cyano-5-(trifluoromethyl)pyrrole, 4-Bromo-2-(p-chlorophenyl)-5-(trifluoromethyl)pyrrole-3-carbonitrile, AC 303268, CL 303268, Econea, Econea 028, Tralopyril
IUPAC-namn 4-bromo-2-(4-chlorophenyl)-5-(trifluoromethyl)-1H-pyrrole-3-carbonitrile
LEDER FC(F)(F)c1[nH]c(c2ccc(Cl)cc2)c(C#N)c1Br
Specifikationer
Kemiskt namn eller material Tralopyril
Kvantitet 50 mg
InChI formel InChI=1S/C12H5BrClF3N2/c13-9-8(5-18)10(19-11(9)12(15,16)17)6-1-3-7(14)4-2-6/h1-4,19H
Formel vikt 347.9277