Läs mer
N-Deethyl Dorzolamide Hydrochloride, TRC
Produktkod. 30362006
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30362006 10 mg
1 options
Produktkod. 30362006 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD22855510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dorzolamide Impurity D
CAS 164455-27-0
Molekylformel C8 H12 N2 O4 S3 . Cl H
Kvantitet 10 mg
Synonym 4H-Thieno[2,3-b]thiopyran-2-sulfonamide, 4-amino-5,6-dihydro-6-methyl-, 7,7-dioxide, monohydrochloride, (4S,6S)- (9CI), Dorzolamide Related Compound D (USP), (4S,6S)-4-Amino-6-methyl-5,6-dihydro-4H-thieno[2,3-b]thiopyran-2-sulfonamide 7,7-Dioxide Hydrochloride, N-Desethyl Dorzolamide HCl
InChI formel InChI=1S/C8H12N2O4S3.ClH/c1-4-2-6(9)5-3-7(17(10,13)14)15-8(5)16(4,11)12;/h3-4,6H,2,9H2,1H3,(H2,10,13,14);1H/t4-,6-;/m0./s1
LEDER Cl.C[C@H]1C[C@H](N)c2cc(sc2S1(=O)=O)S(=O)(=O)N
IUPAC-namn (4S,6S)-4-amino-6-methyl-7,7-dioxo-5,6-dihydro-4H-thieno[2,3-b]thiopyran-2-sulfonamide;hydrochloride
Molekylvikt (g/mol) 332.85
Formel vikt 331.9726