Läs mer
N-Demethyl-N-formyl Clarithromycin, TRC
Produktkod. 30351143
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30351143 5 mg
30355430 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30351143 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2311355MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 127140-69-6
Molekylformel C38 H67 N O14
Molekylvikt (g/mol) 761.94
Synonym N-formyl clarithromycin, 3''-N-Demethyl-3''-N-formyl-6-O-methylerythromycin A
IUPAC-namn N-[(2S,3R,4S,6R)-2-[[(3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-14-ethyl-12,13-dihydroxy-4-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-7-methoxy-3,5,7,9,11,13-hexamethyl-2,10-dioxo-oxacyclotetradec-6-yl]oxy]-3-hydroxy-6-methyloxan-4-yl]-N-methylformamide
LEDER CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C=O)[C@@](C)(C[C@@H](C)C(=O)[C@H](C)[C@@H](O)[C@]1(C)O)OC
Specifikationer
Kemiskt namn eller material N-formyl clarithromycin
Kvantitet 5 mg
InChI formel InChI=1S/C38H67NO14/c1-14-26-38(10,46)31(43)21(4)28(41)19(2)16-37(9,48-13)33(53-35-29(42)25(39(11)18-40)15-20(3)49-35)22(5)30(23(6)34(45)51-26)52-27-17-36(8,47-12)32(44)24(7)50-27/h18-27,29-33,35,42-44,46H,14-17H2,1-13H3/t19-,20-,21+,22+,23-,24+,25+,26-,27+,29-,30+,31-,32+,33-,35+,36-,37-,38-/m1/s1
Formel vikt 761.4562