Läs mer
N-Demethyl Olanzapine, TRC
Produktkod. 17971295
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17971295 100 mg
17931175 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17971295 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD230970100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 161696-76-0
Molekylformel C16 H18 N4 S
Molekylvikt (g/mol) 298.41
Synonym N-Desmethylolanzapine, Desmethylolanzapine, 2-Methyl-4-(1-piperazinyl)-10H-thieno[2,3-b][1,5]benzodiazepine, 4'-Desmethylolanzapine, LY 170055, N-Demethylolanzapine
IUPAC-namn 2-methyl-4-piperazin-1-yl-10H-thieno[2,3-b][1,5]benzodiazepine
LEDER Cc1cc2C(=Nc3ccccc3Nc2s1)N4CCNCC4
Specifikationer
Kemiskt namn eller material Desmethylolanzapine
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C16H18N4S/c1-11-10-12-15(20-8-6-17-7-9-20)18-13-4-2-3-5-14(13)19-16(12)21-11/h2-5,10,17,19H,6-9H2,1H3
Formel vikt 298.1252
Procent renhet >95%