missing translation for 'onlineSavingsMsg'
Learn More

N-Demethyl Trimebutine-d5 Hydrochloride, TRC

Product Code. 30354767
Change view
Click to view available options
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30354767 10 mg
30365610 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30354767 Supplier Toronto Research Chemicals Supplier No. TRCD23134210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1286632-72-1
Molekylformel C21 H22 D5 N O5 . H Cl
Molekylvikt (g/mol) 414.93
LEDER Cl.[2H]C([2H])([2H])C([2H])([2H])C(COC(=O)c1cc(OC)c(OC)c(OC)c1)(NC)c2ccccc2

Specifications

Kemiskt namn eller material N-Demethyl Trimebutine-d5 hydrochloride
Kvantitet 10 mg
InChI formel InChI=1S/C21H27NO5.ClH/c1-6-21(22-2,16-10-8-7-9-11-16)14-27-20(23)15-12-17(24-3)19(26-5)18(13-15)25-4;/h7-13,22H,6,14H2,1-5H3;1H/i1D3,6D2;
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.