Läs mer
N-Demethylclindamycin, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-Desmethylclindamycin |
| CAS | 22431-45-4 |
| Molekylformel | C17 H31 Cl N2 O5 S |
| Kvantitet | 1 mg |
| Synonym | L-threo-α-D-galacto-Octopyranoside, methyl 7-chloro-6,7,8-trideoxy-6-[[[(2S,4R)-4-propyl-2-pyrrolidinyl]carbonyl]amino]-1-thio- (9CI), L-threo-D-galacto-Octopyranoside, methyl 7-chloro-6,7,8-trideoxy-6-(4-propyl-L-2-pyrrolidinecarboxamido)-1-thio-, trans- α- (8CI), L-threo-α-D-galacto-Octopyranoside, methyl 7-chloro-6,7,8-trideoxy-6-[[(4-propyl-2-pyrrolidinyl)carbonyl]amino]-1-thio-, (2S-trans)-, (7S)-7-Chloro-7-deoxy-1'-demethyllincomycin, 1'-Demethylclindamycin, 7-Chloro-N-demethyllincomycin, N-Demethylclindamycin, U 26727A |
| InChI formel | InChI=1S/C17H31ClN2O5S/c1-4-5-9-6-10(19-7-9)16(24)20-11(8(2)18)15-13(22)12(21)14(23)17(25-15)26-3/h8-15,17,19,21-23H,4-7H2,1-3H3,(H,20,24)/t8-,9+,10-,11+,12-,13+,14+,15+,17+/m0/s1 |
| LEDER | CCC[C@H]1CN[C@@H](C1)C(=O)N[C@H]([C@H](C)Cl)[C@H]2O[C@H](SC)[C@H](O)[C@@H](O)[C@H]2O |
| IUPAC-namn | (2S,4R)-N-[(1S,2S)-2-chloro-1-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-4-propylpyrrolidine-2-carboxamide |
| Molekylvikt (g/mol) | 410.96 |
| Formel vikt | 410.1642 |