Läs mer
N-Demethylivabradine, TRC
Produktkod. 30351284
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30351284 10 mg
1 options
Produktkod. 30351284 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29299510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Demethylivabradine
CAS 215935-23-2
Molekylformel C26H34N2O5
Kvantitet 10 mg
Synonym 3-[3-[[[(7S)-3,4-Dimethoxybicyclo[4.2.0]octa-1,3,5-trien-7-yl]methyl]amino]propyl]-1,3,4,5-tetrahydro-7,8-dimethoxy-2H-3-benzazepin-2-one, N-Demethylivabradine, S 18982
InChI formel InChI=1S/C26H34N2O5/c1-30-22-11-17-6-9-28(26(29)14-18(17)12-23(22)31-2)8-5-7-27-16-20-10-19-13-24(32-3)25(33-4)15-21(19)20/h11-13,15,20,27H,5-10,14,16H2,1-4H3/t20-/m1/s1
LEDER C(NCCCN1C(=O)CC=2C(CC1)=CC(OC)=C(OC)C2)[C@@H]3C=4C(C3)=CC(OC)=C(OC)C4
Molekylvikt (g/mol) 454.56