Läs mer
N-Des[2-(1-methyl-2-pyrrolidinyl)ethyl] Udenafil, TRC
Produktkod. 30350051
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30350051 250 mg
30353989 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30350051 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD292655250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 319491-68-4
Molekylformel C18 H23 N5 O4 S
Molekylvikt (g/mol) 405.4713
Synonym Benzenesulfonamide, 3-(4,7-dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-propoxy- (9CI), 3-(6,7-Dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-propoxybenzenesulfonamide, DA 8164, Udenafil N-desalkyl
IUPAC-namn 3-(1-methyl-7-oxo-3-propyl-4H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-propoxybenzenesulfonamide
LEDER CCCOc1ccc(cc1C2=NC(=O)c3c(N2)c(CCC)nn3C)S(=O)(=O)N
Specifikationer
Kemiskt namn eller material N-Desalkyludenafil
Kvantitet 250 mg
InChI formel InChI=1S/C18H23N5O4S/c1-4-6-13-15-16(23(3)22-13)18(24)21-17(20-15)12-10-11(28(19,25)26)7-8-14(12)27-9-5-2/h7-8,10H,4-6,9H2,1-3H3,(H2,19,25,26)(H,20,21,24)
Formel vikt 405.1471