missing translation for 'onlineSavingsMsg'
Läs mer

N-des(5-methyl-1,3,4-oxadiazol-2-yl)carbonyl-N-Cbz-Raltegravir, TRC

Produktkod. 30335040
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30335040 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30335040 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB2765905MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material Benzyl [1-[4-[((4-Fluorobenzyl)amino)carbonyl]-5-hydroxy-1-methyl-6-oxo-1,6-dihydropyrimidin-2-yl]-1-methylethyl]carbamate
CAS 518048-02-7
Molekylformel C24 H25 F N4 O5
Kvantitet 5 mg
Synonym Benzyl [1-[4-[[(4-Fluorobenzyl)amino]carbonyl]-5-hydroxy-1-methyl-6-oxo-1,6-dihydropyrimidin-2-yl]-1-methylethyl]carbamate, Carbamic acid, N-[1-[4-[[[(4-fluorophenyl)methyl]amino]carbonyl]-1,6-dihydro-5-hydroxy-1-methyl-6-oxo-2-pyrimidinyl]-1-methylethyl]-, phenylmethyl ester, Carbamic acid, [1-[4-[[[(4-fluorophenyl)methyl]amino]carbonyl]-1,6-dihydro-5-hydroxy-1-methyl-6-oxo-2-pyrimidinyl]-1-methylethyl]-, phenylmethyl ester (9CI), Benzyl [1-[4-[[(4-fluorobenzyl)amino]carbonyl]-5-hydroxy-1-methyl-6-oxo-1,6-dihydropyrimidin-2-yl]-1-methylethyl]carbamate, [1-[4-(4-Fluorobenzylcarbamoyl)-5-hydroxy-1-methyl-6-oxo-1,6-dihydropyrimidin-2-yl]-1-methylethyl]carbamic acid benzyl ester, Benzyl N-[2-[4-[(4-Fluorobenzyl)carbamoyl]-5-hydroxy-1-methyl-6-oxo-1,6-dihydropyrimidin-2-yl]propan-2-yl]carbamate
InChI formel InChI=1S/C24H25FN4O5/c1-24(2,28-23(33)34-14-16-7-5-4-6-8-16)22-27-18(19(30)21(32)29(22)3)20(31)26-13-15-9-11-17(25)12-10-15/h4-12,30H,13-14H2,1-3H3,(H,26,31)(H,28,33)
LEDER CN1C(=O)C(=C(N=C1C(C)(C)NC(=O)OCc2ccccc2)C(=O)NCc3ccc(F)cc3)O
IUPAC-namn benzyl N-[2-[4-[(4-fluorophenyl)methylcarbamoyl]-5-hydroxy-1-methyl-6-oxopyrimidin-2-yl]propan-2-yl]carbamate
Molekylvikt (g/mol) 468.48
Formel vikt 468.1809
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.