Läs mer
N-des(5-methyl-1,3,4-oxadiazol-2-yl)carbonyl-N-Cbz-Raltegravir, TRC
Produktkod. 30331685
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30331685 10 mg
1 options
Produktkod. 30331685 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB27659010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Benzyl [1-[4-[((4-Fluorobenzyl)amino)carbonyl]-5-hydroxy-1-methyl-6-oxo-1,6-dihydropyrimidin-2-yl]-1-methylethyl]carbamate
CAS 518048-02-7
Molekylformel C24 H25 F N4 O5
Kvantitet 10 mg
Synonym Benzyl [1-[4-[[(4-Fluorobenzyl)amino]carbonyl]-5-hydroxy-1-methyl-6-oxo-1,6-dihydropyrimidin-2-yl]-1-methylethyl]carbamate, Carbamic acid, N-[1-[4-[[[(4-fluorophenyl)methyl]amino]carbonyl]-1,6-dihydro-5-hydroxy-1-methyl-6-oxo-2-pyrimidinyl]-1-methylethyl]-, phenylmethyl ester, Carbamic acid, [1-[4-[[[(4-fluorophenyl)methyl]amino]carbonyl]-1,6-dihydro-5-hydroxy-1-methyl-6-oxo-2-pyrimidinyl]-1-methylethyl]-, phenylmethyl ester (9CI), Benzyl [1-[4-[[(4-fluorobenzyl)amino]carbonyl]-5-hydroxy-1-methyl-6-oxo-1,6-dihydropyrimidin-2-yl]-1-methylethyl]carbamate, [1-[4-(4-Fluorobenzylcarbamoyl)-5-hydroxy-1-methyl-6-oxo-1,6-dihydropyrimidin-2-yl]-1-methylethyl]carbamic acid benzyl ester, Benzyl N-[2-[4-[(4-Fluorobenzyl)carbamoyl]-5-hydroxy-1-methyl-6-oxo-1,6-dihydropyrimidin-2-yl]propan-2-yl]carbamate
InChI formel InChI=1S/C24H25FN4O5/c1-24(2,28-23(33)34-14-16-7-5-4-6-8-16)22-27-18(19(30)21(32)29(22)3)20(31)26-13-15-9-11-17(25)12-10-15/h4-12,30H,13-14H2,1-3H3,(H,26,31)(H,28,33)
LEDER CN1C(=O)C(=C(N=C1C(C)(C)NC(=O)OCc2ccccc2)C(=O)NCc3ccc(F)cc3)O
IUPAC-namn benzyl N-[2-[4-[(4-fluorophenyl)methylcarbamoyl]-5-hydroxy-1-methyl-6-oxopyrimidin-2-yl]propan-2-yl]carbamate
Molekylvikt (g/mol) 468.48
Formel vikt 468.1809